Learn More
Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Boots, Ties
GREENER_CHOICE
Catalog Number p-4365030
4479.67 SEK valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
Small/Medium
Sterile cleanroom boots with anti-slip soles feature innovative CLEAN-DON™ technology for aseptic donning. Suitable for EU GMP ISO 5 Grade A sterile cleanrooms.
  • Sterile cleanroom boots with ties, vinyl
  • Average bacterial filtration efficiency (BFE) of 96%
  • Made of 100% polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-Don ties and 2-inch edge vinyl, for an enhance moisture barrier
  • Made without latex, silicone or the BHT preservative and are designed for single-use
  • 100 Pairs/200 Each
  • CE certified (CE Simple)
  • Each individually bagged pair of boot covers has a sterilization indicator as a visual that it's undergone gamma sterilization
  • A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Certifications
CE Marked, Category III, PPE
TRUSTED_SUSTAINABILITY
Size Small/Medium
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Top with Ankle Ties
Packaging Quantity 100 Pairs/Cs.
Packaging Type Triple Bagged, Vacuum Packed
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)