Learn More
Kimberly-Clark Professional™ Purple Nitrile-XTRA™ Gloves
Kimberly-Clark™ Professional Purple Nitrile-XTRA™ Gloves are made of a highly durable nitrile polymer and are powder free.
168.00 SEK
Specifications
Certifications/Compliance ASTM D6319, FDA 510(k)
Coating Material Textured fingertips
Color Purple
Disposable Yes
Glove Cuff Style Beaded/Rolled
View More Specs
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11819620 View Documents Kimberly-Clark Professional™
97610
X-Small 168.00 SEK
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment
06-08-2019
Add to basket
11839610 View Documents Kimberly-Clark Professional™
97611
Small 168.00 SEK
Pack of 50 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 25
Add to basket
11829620 View Documents Kimberly-Clark Professional™
97612
Medium 168.00 SEK
Pack of 50 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 71
Add to basket
11839620 View Documents Kimberly-Clark Professional™
97613
Large 168.00 SEK
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment
29-07-2019
Add to basket
11849620 View Documents Kimberly-Clark Professional™
97614
X-Large 168.00 SEK
Pack of 50 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 14
Add to basket
Description
Description
  • Guard against incidental splashes
  • Not made with natural rubber latex (latex free)
  • Ambidextrous
  • Beaded cuff prevent roll-down and allow for easy donning
  • Excellent dexterity and tactile sensitivity, thanks to the textured fingertips
  • Cleared for use in chemotherapy.
  • Fully recyclable through the RightCycle program (by Kimberly-Clark Professional), so lab gloves won’t end up in landfills
Certifications
510K approved for medical use
Compliance
Meets ASTM™ D6319 standards; FDA 510(k) approved for medical use.
Specifications
Specifications
ASTM D6319, FDA 510(k)
Purple
Beaded/Rolled
Yes
21
Acrylonitrile Butadiene
Textured fingertips
Yes
Ambidextrous
Powder-free
30.5cm