Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Brand: DWK Life Sciences 60811B612
Additional Details : Weight : 0.01000kg
Description
- Small case quantities
- Manufactured from clear 33 expansion, borosilicate glass
- Attached closed top black phenolic closures
Specifications
A | |
Clear | |
Borosilicate Glass | |
Screw | |
Phenolic | |
70 mm |
PTFE/Rubber | |
28 mm | |
25 mL | |
6.76 dr. | |
Screw Cap (Closed-top) |