Learn More
DWK Life Sciences Kimble™ KIMAX™ Colorware™ Flasks
Colorful enamel graduations enhance readability and indicate approximate volumes
1009.19 SEK - 1170.00 SEK
Specifications
Closure Material | Rubber |
---|---|
Material | Borosilicate Glass |
Writing Surface | Matte Finish Marking |
Mouth Style | Narrow |
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
11733839
|
DWK Life Sciences
26500G-250 |
250 mL | Cool Green Ceramic Enamel scale |
1170.00 SEK
Pack of 12 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
11812330
|
DWK Life Sciences
26500R-250 |
250 mL | Raging Red Ceramic Enamel scale graduations |
1170.00 SEK
Pack of 12 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
11748083
|
DWK Life Sciences
26500Y250 |
250 mL | Sunny Yellow Ceramic Enamel scale graduations |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11707953
|
DWK Life Sciences
26500Y50 |
50 mL | Sunny Yellow Ceramic Enamel scale graduations |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11702464
|
DWK Life Sciences
26500B250 |
250 mL | Bright Blue Ceramic Enamel scale |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Colorful enamel graduations enhance readability and indicate approximate volumes
- Narrow mouth
- Durable, matte-finish marking spot provides good writing surface
- Without stoppers
Compliance
Designed from ASTM™ E 1404, Type I, Class 1 requirements
Specifications
Rubber | |
Borosilicate Glass | |
Matte Finish Marking | |
Narrow |