Learn More
DWK Life Sciences Kimble™ Kontes™ 90° Hose Inlet Adapter
Fits 0.375 in. ID tubing
Brand: DWK Life Sciences 1830002440
Additional Details : Weight : 0.80000kg
Description
- standard taper 24/40
- Hose connection No. 2
- H: 80mm
Specifications
0.37 in. | |
3.14 in. | |
Hose connection 2 | |
24/40 |
9.39mm | |
80mm | |
90° Inlet Adapter |