Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Three-way Luer Stopcock
Heavy-duty nylon
Brand: DWK Life Sciences 420163-4503
Additional Details : Weight : 1.81440kg
Description
Heavy-duty nylon
- Three-way control with locking fittings on all ports
Specifications
3-way Stopcock | |
FlexColumn™ | |
HDPE | |
Female Luer to Male Luer |
Nylon | |
Locking fittings on all ports | |
Female Luer to Male Luer |