Learn More
Alfa Aesar™ Kinase buffer III (5X)
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J61566.AK
Additional Details : Weight : 0.00025kg
 View more versions of this product
Product Code. 15494149
  • 470.50 SEK / 250mL
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
  • Formulation: 100mM PIPES and 50mM manganese chloride at pH 8.0
Specifications
Specifications
Kinase buffer III (5X)
Liquid
100mM PIPES and 50mM manganese chloride at pH 8.0
250mL