missing translation for 'onlineSavingsMsg'
Learn More

Kir2.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14135-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882634

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inward rectifying potassium channel that is activated by phosphatidylinositol 4,5-bisphosphate and that probably participates in controlling the resting membrane potential in electrically excitable cells. Probably participates in establishing action potential waveform and excitability of neuronal and muscle tissues. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
Specifications

Specifications

Kir2.2
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52187, P52188, Q14500
hIRK, hIRK1, hkir2.2x, IRK 2, IRK2, KCNJ12, kcnj12x, KCNJN1, Kir2.2, Kir2.2v
Rabbit
IgG
150 μL
-20°C
Primary
117052, 16515, 3768
Flow Cytometry, Western Blot
Unconjugated
KCNJ12
Liquid
KCNJ12
Kir2.2 Fusion Protein Ag5349
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers