missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Kir3.2 (KCNJ6) Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA518802
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins and may be involved in the regulation of insulin secretion by glucose. It associates with two other G-protein-activated potassium channels to form a heteromultimeric pore-forming complex.
Specifica
| Kir3.2 (KCNJ6) | |
| Polyclonal | |
| Unconjugated | |
| KCNJ6 | |
| BIR1; G protein-activated inward rectifier potassium channel 2; Girk2; GIRK-2; hiGIRK2; Inward rectifier K(+) channel Kir3.2; inward rectifier potassium channel KIR3.2; KATP2; KATP-2; KCNJ6; Kcnj7; KIR3.2; KPLBS; MGC126596; Potassium channel, inwardly rectifying subfamily J member 6; potassium channel, inwardly rectifying subfamily J, member 6; potassium inwardly-rectifying channel J6; potassium inwardly-rectifying channel, subfamily J, member 6; potassium voltage-gated channel subfamily J member 6; W; weaver; wv | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3763 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P48051 | |
| KCNJ6 | |
| Synthetic peptide sequence (SSKLNQHAELET) corresponding to the C-terminus amino acids of KCNJ6 (aa 386-398). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto