missing translation for 'onlineSavingsMsg'
Learn More

KIR3DL3/CD158z Antibody (1136B), PE/Cy5.5, Novus Biologicals™

Product Code. 30026289 Shop All Bio Techne Products
Change view
Click to view available options
Kvantitet:
0.1 mL
Unit Size:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30026289 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30026289 Supplier Novus Biologicals Supplier No. FAB8919PECY55

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DL3/CD158z
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona 1136B
Konjugera PE-Cyanine5.5
Utspädning Flow Cytometry
Formulering PBS
Gene Alias CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1
Värd art Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z, Met1-Leu322, Accession # Q8N743
Reningsmetod Protein A or G purified from cell culture supernatant
Kvantitet 0.1 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 115653
Målarter Human
Innehåll och lagring Store at 4C in the dark. Do not freeze.
Form Purified
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.