missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DX1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17583-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
KIR3DX1 | |
Polyclonal | |
Unconjugated | |
KIR3DX1 | |
KIR3DL0, KIR3DX1, LENG12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90011 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H7L2 | |
KIR3DX1 | |
KIR3DX1 Fusion Protein Ag11220 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |