Läs mer
KIRA6, MedChemExpress
Produktkod. 16460318
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16460318 5 mg
16441849 1 mg
16427858 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16460318 Leverantör Medchem Express Leverantörsnummer HY19708
MedChemExpress KIRA6 is an advanced small-molecule IRE1α RNase kinase inhibitor with an IC50 of 0.6 μM. KIRA6 can trigger an apoptotic response.
KIRA6 is an advanced small-molecule IRE1α RNase kinase inhibitor with an IC50 of 0.6 μM. KIRA6 can trigger an apoptotic response.
Specifikationer
Kemiskt namn eller material KIRA6
CAS 1589527-65-0
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C28H25F3N6O
Kvantitet 5 mg
Löslighetsinformation DMSO : 25 mg/mL (48.21 mM; Need ultrasonic)
LEDER O=C(NC1=CC=CC(C(F)(F)F)=C1)NC2=C3C=CC=CC3=C(C4=C5C(N)=NC=CN5C(C(C)(C)C)=N4)C=C2
Molekylvikt (g/mol) 518.53
Formel vikt 518.53
Procent renhet 98.02%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease