Läs mer
Kitol Diacetate (Mixture of Diastereomers), TRC
Produktkod. 17993825
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17993825 50 mg
17923075 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17993825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK57050550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13828-13-2
Molekylformel C44 H64 O4
Molekylvikt (g/mol) 656.9766
Synonym 3-Cyclohexene-1,2-dimethanol, 3,6-dimethyl-5-[2-methyl-4-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3-butadienyl]-6-[4-methyl-6-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3,5-hexatrienyl]-, diacetate (9CI), 3-Cyclohexene-1,2-dimethanol, 3,6-dimethyl-5-[2-methyl-4-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3-butadienyl]-6-[4-methyl-6-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3,5-hexatrienyl]-, diacetate, stereoisomer (8CI), (3,6-Dimethyl-5-(2-methyl-4-(2,6,6-trimethylcyclohex-1-en-1-yl)buta-1,3-dien-1-yl)-6-(4-methyl-6-(2,6,6-trimethylcyclohex-1-en-1-yl)hexa-1,3,5-trien-1-yl)cyclohex-3-ene-1,2-diyl)bis(methylene) diacetate, Kitol 2, Retinol acetate dimer, Retinyl acetate dimer, [6-(Acetoxymethyl)-2,5-dimethyl-4-[2-methyl-4-(2,6,6-trimethylcyclohexen-1-yl)buta-1,3-dienyl]-5-[4-methyl-6-(2,6,6-trimethylcyclohexen-1-yl)hexa-1,3,5-trienyl]cyclohex-2-en-1-yl]methyl acetate, 3,6-Dimethyl-5-[2-methyl-4-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3-butadienyl]-6-[4-methyl-6-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,3,5-hexatrienyl]-3-cyclohexene-1,2-dimethano Diacetate Stereoisomer
IUPAC-namn [6-(acetyloxymethyl)-2,5-dimethyl-4-[2-methyl-4-(2,6,6-trimethylcyclohexen-1-yl)buta-1,3-dienyl]-5-[4-methyl-6-(2,6,6-trimethylcyclohexen-1-yl)hexa-1,3,5-trienyl]cyclohex-2-en-1-yl]methyl acetate
LEDER CC(=O)OCC1C(COC(=O)C)C(C)(C=CC=C(C)C=CC2=C(C)CCCC2(C)C)C(C=C(C)C=CC3=C(C)CCCC3(C)C)C=C1C
Specifikationer
Kemiskt namn eller material Kitol Diacetate
Kvantitet 50 mg
InChI formel InChI=1S/C44H64O4/c1-30(19-21-39-32(3)17-14-23-42(39,8)9)16-13-25-44(12)37(26-31(2)20-22-40-33(4)18-15-24-43(40,10)11)27-34(5)38(28-47-35(6)45)41(44)29-48-36(7)46/h13,16,19-22,25-27,37-38,41H,14-15,17-18,23-24,28-29H2,1-12H3
Formel vikt 656.4805