Learn More
Ecolab™ Klerwipe™ Silicone
Sterile low particulate wipes impregnated with silicone and an IPA carrier
Brand: Ecolab™ 3079440
Additional Details : CAS Number : 67-63-0 Weight : 0.50000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- The blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
- A sterile ready-to-use maintenance wipe in a resealable pouch, featuring Klertear on outer wraps, for small surfaces.
Specifications
Sterile Silicone Pouch Wipe | |
200 x 200 mm | |
20 x 15 Wipes |