missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11881-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is a rabbit polyclonal antibody raised against a full-length human KLF10 protein and specifically recognizes the 52 kDa KLF10 protein.
This gene encodes a member of a family of proteins that feature C2H2-type zinc finger domains. The encoded protein is a transcriptional repressor that acts as an effector of transforming growth factor beta signaling. Activity of this protein may inhibit the growth of cancers, particularly pancreatic cancer. Alternative splicing results in multiple transcript variants.Specifications
KLF10 | |
Polyclonal | |
Unconjugated | |
KLF10 | |
EGR alpha, EGRA, KLF10, Krueppel like factor 10, Kruppel like factor 10, TIEG, TIEG 1, TIEG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7071 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13118 | |
KLF10 | |
KLF10 Fusion Protein Ag2482 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |