missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF17 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17612-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5′-CACCC-3′ sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.Specifications
KLF17 | |
Polyclonal | |
Unconjugated | |
KLF17 | |
KLF17, Krueppel like factor 17, Kruppel like factor 17, Zfp393, Zinc finger protein 393, ZNF393 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
128209 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5JT82 | |
KLF17 | |
KLF17 Fusion Protein Ag11776 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |