missing translation for 'onlineSavingsMsg'
Learn More

KLF17 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17612-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877544

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5′-CACCC-3′ sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Specifications

Specifications

KLF17
Polyclonal
Unconjugated
KLF17
KLF17, Krueppel like factor 17, Kruppel like factor 17, Zfp393, Zinc finger protein 393, ZNF393
Rabbit
Antigen Affinity Chromatography
RUO
128209
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5JT82
KLF17
KLF17 Fusion Protein Ag11776
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers