Learn More
KLF6 Mouse anti-Human, Unconjugated, Clone: 2F8B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67297-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839845
  • 3665.00 SEK / 150µL
Estimated Shipment: 29-11-2022
to see stock.
Description
Description
The KLF6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLF6. This antibody recognizes human, mouse antigen. The KLF6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KLF6
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Human
ELISA, Western Blot
2F8B3
Unconjugated
150 μL
1316
IgG1