missing translation for 'onlineSavingsMsg'
Learn More

KLF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14716-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823504

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody raise against the full length of KLF6 gene of human origin, and can recognize both of KLF6, included KLF6 (∽32 kDa), p-KLF6(∽35-46 kDa). Otherwise, there may be a non-specific band(50 kDa) shown.

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.
Specifications

Specifications

KLF6
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08584, O35819, Q99612
B cell derived protein 1, COPEB, CPBP, GBF, KLF6, Krueppel like factor 6, Kruppel like factor 6, PAC1, Proto oncogene BCD1, ST12, Transcription factor Zf9, ZF9
Rabbit
IgG
20 μL
-20°C
Primary
1316, 23849, 58954
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
KLF6
Liquid
Klf6
KLF6 Fusion Protein Ag6440
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers