missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KLHDC1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825535
|
Proteintech
27277-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815535
|
Proteintech
27277-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.Specifications
KLHDC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80YG3, Q8N7A1 | |
122773, 271005, 314190 | |
KLHDC1 Fusion Protein Ag23704 | |
Primary | |
-20°C | |
KLHDC1 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
kelch domain containing 1, KLHDC1, MST025 | |
Klhdc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |