missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KLHL14 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896404
|
Proteintech
16693-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16886404
|
Proteintech
16693-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KLHL14 is a member of the KLHL family.Specifications
KLHL14 | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q69ZK5, Q9P2G3 | |
225266, 364823, 57565 | |
KLHL14 Fusion Protein Ag10065 | |
Primary | |
-20°C | |
KLHL14 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
kelch like 14 (Drosophila), Kelch like protein 14, KIAA1384, KLHL14 | |
Klhl14 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |