missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16951-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Probable substrate-specific adapter of an E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.Specifications
KLHL3 | |
Polyclonal | |
Unconjugated | |
KLHL3 | |
kelch like 3 (Drosophila), Kelch like protein 3, KIAA1129, KLHL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100503085, 26249, 498697 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E0CZ16, F1LZ52, Q9UH77 | |
Klhl3 | |
KLHL3 Fusion Protein Ag10580 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |