missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KLHL9 |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876855
|
Proteintech
55394-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16866855
|
Proteintech
55394-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KLHL9 is substrate-specific adapter for a CUL3-based E3 ubiquitin-protein ligase complex. Within this complex, it controls the dynamic behavior of AURKB on mitotic chromosomes and thereby coordinates faithful mitotic progression and completion of cytokinesis.Specifications
KLHL9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9P2J3 | |
55958 | |
Peptide | |
Primary | |
-20°C | |
KLHL9 |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
kelch like 9 (Drosophila), Kelch like protein 9, KIAA1354, KLHL9 | |
KLHL9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |