Learn More
KLRB1/CD161 Mouse anti-Human, Unconjugated, Clone: 3D8F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67537-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The KLRB1/CD161 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLRB1/CD161. This antibody recognizes human antigen. The KLRB1/CD161 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
KLRB1/CD161 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
3D8F5 | |
Unconjugated | |
150 μL | |
3820 | |
IgG1 |