Learn More
KLRB1/CD161 Mouse anti-Human, Unconjugated, Clone: 3D8F5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67537-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16830226
  • 4721.67 SEK / 150µL
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
The KLRB1/CD161 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLRB1/CD161. This antibody recognizes human antigen. The KLRB1/CD161 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
KLRB1/CD161
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
3D8F5
Unconjugated
150 μL
3820
IgG1