Läs mer
Tocris Bioscience™ KML 29
GREENER_CHOICE
Produktkod. 30002040
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30002040 50 mg
30002032 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30002040 Leverantör Tocris Bioscience™ Leverantörsnummer 4872/50
Highly potent and selective MAGL inhibitor
Highly selective and potent monoacylglycerol lipase (MAGL) inhibitor. Exhibits potent inhibition of human, mouse and rat MAGL (IC50 values are 5.9, 15 and 43 nM, respectively). Exhibits no detectable inhibition of FAAH (IC50 > 50000 nM). Potently and selectively blocks hydrolysis of 2-arachidonoylglycerol (2-AG) in mice (IC50 = 2.5 nM and >50 μM for 2-AG and AEA respectively).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1380424-42-9
Molekylformel C24H21F6NO7
Specifikationer
Kemiskt namn eller material KML 29
Kvantitet 50 mg
Formel vikt 549.42
Molekylvikt (g/mol) 549.422