Läs mer
KN-93 hydrochloride, MedChemExpress
Produktkod. 16482637
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16482637 50 mg
16479127 5 mg
16463476 25 mg
16456548 10 mg
16401529 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16482637 Leverantör Medchem Express Leverantörsnummer HY15465A
MedChemExpress KN-93 hydrochloride is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM.
KN-93 hydrochloride is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM.
Specifikationer
Kemiskt namn eller material KN-93 hydrochloride
CAS 1956426-56-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C26H30Cl2N2O4S
Kvantitet 50 mg
Löslighetsinformation DMSO : ≥ 31 mg/mL (57.67 mM) ∣H2O : 0.45 mg/mL (0.84 mM; Need ultrasonic and warming)
LEDER [H]Cl.O=S(C1=CC=C(OC)C=C1)(N(C2=CC=CC=C2CN(C/C=C/C3=CC=C(Cl)C=C3)C)CCO)=O
Molekylvikt (g/mol) 537.5
Formel vikt 537.5
Procent renhet 99.92%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease