missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
KN-93 phosphate, MedChemExpress
Beskrivning
KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II, with Ki of 370 nM.
Specifikationer
Specifikationer
| Kemiskt namn eller material | KN-93 phosphate |
| CAS | 1913269-12-1 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C26H32ClN2O8PS |
| Kvantitet | 50 mg |
| Löslighetsinformation | DMSO : 100 mg/mL (166.94 mM; Need ultrasonic) |
| LEDER | OP(O)(O)=O.O=S(C1=CC=C(OC)C=C1)(N(C2=CC=CC=C2CN(C/C=C/C3=CC=C(Cl)C=C3)C)CCO)=O |
| Molekylvikt (g/mol) | 599.03 |
| Formel vikt | 599.03 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?