Läs mer
Tocris Bioscience™ KN 93 phosphate
Beskrivning
KN 93 phosphate is a CaM kinase II inhibitor; water soluble version of KN 93 (Cat. No. 1278).
Specifikationer
Specifikationer
| Kvantitet | 1 mg |
| Produkttyp | KN 93 phosphate |
| Renhet | >98% |
| Molekylvikt (g/mol) | 599.03 |
| Inhibitorer | KN 93 phosphate |
For Research Use Only