Läs mer
KO-947, MedChemExpress
Produktkod. 16493146
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16493146 25 mg -
16487307 5 mg 5mg
16434598 10 mg -
16430897 50 mg -
16425446 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16493146 Leverantör Medchem Express Leverantörsnummer HY112181
MedChemExpress KO-947 is a potent and selective inhibitor of ERK1/2 kinases with potential utility in MAPK pathway dysregulated tumors.
KO-947 is a potent and selective inhibitor of ERK1/2 kinases with potential utility in MAPK pathway dysregulated tumors.
Specifikationer
Kemiskt namn eller material KO-947
Fysisk form Solid
CAS 1695533-89-1
Färg Off-White
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H17N5O
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 62.5 mg/mL (175.86 mM; Need ultrasonic)
LEDER O=C1NC2=CC(NN=C3C4=CC=NC=C4)=C3C=C2CN1CC5=CC=CC=C5
Molekylvikt (g/mol) 355.39
Formel vikt 355.39
Procent renhet 98.08%