Läs mer
MEDICOM Kolmi™ Feeldry Type IIR Tie-On Mask
Produktkod. 31781379 Handla allt Medicom Produkter
Change view
Färg:
Blue
Green
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Färg
31781379 Green
31781289 White
31781288 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 31781379 Leverantör MEDICOM Leverantörsnummer M3420140
Kolmi™ Feeldry Type IIR mask: protects others and wearer from biological fluid splashes with durable comfort.
  • Protects the environment from droplets emitted by the wearer while providing protection from splashes of biological fluids
  • Effective against bacteria, particles and viruses (BFE, PFE, and VFE)
  • Kolmi masks feature the exclusive Origami folding for greater comfort and an ideal fit on the wearer's face
  • Featuring a wide and ultra-flexible nose strip, they ensure an excellent fit to the face and effectively limit leakage around the nose, thereby significantly reducing fogging
  • For maximum comfort, the inner layer is designed with a material that limits irritation to the face
  • With Feeldry technology the inner layer has been specially designed to absorb perspiration and avoid the feeling of humidity inside the mask
  • Hypoallergenic
  • NEW PACKAGING : Each tie-on is folded under each mask for convenient and tidy single removal
Specifikationer
Produkttyp Medical/Procedural Face Mask
Färg Green
Fästtyp Ties
Material Polypropylene
Storlek One Size
Sterilitet Non-sterile
Stil Pleated