Learn More
DWK Life Sciences Kimble™ Kontes™ Chromaflex Chromatography Fittings : Adapter with 1/2-20 to 1/4-28 Thread
Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads.
Brand: DWK Life Sciences 420804-0001
Additional Details : Weight : 0.01000kg
Description
Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads.
- CTFE Threaded Adapter with 1/2-20 to 1/4-28 Thread
- Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads
- Case of 2
Specifications
Adapter | |
1 | |
Glass |
Chromatography | |
Kimble Kontes Hi-Vac PTFE plug |