Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
Brand: DWK Life Sciences 774240-0019
Additional Details : Weight : 0.15300kg
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
- Leak-proof, air-tight seals
- Wide top flange for ease of use
Specifications
Stopper | |
Polyethylene | |
19/22 ,19/38 |