missing translation for 'onlineSavingsMsg'
Learn More

KRBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10386-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826743

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

KRBP
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R1D1, Q9BUY5
K RBP, KRBP, zinc finger protein 426, ZNF426
Rabbit
IgG
20 μL
-20°C
Primary
235028, 79088
Western Blot
Unconjugated
ZNF426
Liquid
ZNF426
KRBP Fusion Protein Ag0585
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers