Läs mer
Tocris Bioscience™ KRIBB11
HSF1 inhibitor
Brand: Tocris Bioscience™ 5480/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
KRIBB11 is a heat shock factor (HSF) inhibitor (IC50 = 1.2 μM). Increases apoptosis in cancer cells treated with the HSP90 inhibitors Geldanamycin (Cat. No. 1368) and 17-AAG (Cat. No. 1515). Blocks downstream induction of HSP27 and HSP70. Inhibits tumor growth in vivo.Specifikationer
| KRIBB11 | |
| N2-1H-Indazol-5-yl-N6-methyl-3-nitro-2,6-pyridinediamine | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 342639-96-7 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use