missing translation for 'onlineSavingsMsg'
Learn More

KRT23 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24049-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16872635

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications

Specifications

KRT23
Polyclonal
Unconjugated
KRT23
CK 23, CK23, Cytokeratin 23, HAIK1, K23, Keratin 23, KRT23
Rabbit
Antigen Affinity Chromatography
RUO
25984
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9C075
KRT23
KRT23 Fusion Protein Ag21129
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers