missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT33B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11048-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B.Specifications
KRT33B | |
Polyclonal | |
Unconjugated | |
KRT33B | |
Ha 3II, HA3II, Hair keratin, type I Ha3 II, hHa3 II, HKA3B, K33B, keratin 33B, KRT33B, KRTHA3A, KRTHA3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3884 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14525 | |
KRT33B | |
KRT33B Fusion Protein Ag1522 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |