missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRT5 (Human) Recombinant Protein
Human KRT5 full-length ORF ( AAH24292, 1 a.a. - 590 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003852-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSRQSSVSFRSGGSRSFSTASAITPSVSRTSFTSVSRSGGGGGGGFGRVSLAGACGVGGYGSRSLYNLGGSKRISISTSGGSFRNRFGAGAGGGYGFGGGAGSGFGFGGGAGGGFGLGGGAGFGGGFGGPGFPVCPPGGIQEVTVNQSLLTPLNLQIDPSIQRVRTEEREQIKTLNNKFASFIDKVRFLEQQNKVLDTKWTLLQEQGTKTVRQNLEPLFEQYINNLRRQLDSIVGERGRLDSELRNMQDLVEDFKNKYEDEINKRTTAENEFVMLKKDVDAAYMNKVELEAKVDALMDEINFMKMFFDAELSQMQTHVSDTSVVLSMDNNRNLDLDSIIAEVKAQYEEIANRSRTEAESWYQTKYEELQQTAGRHGDDLRNTKHEISEMNRMIQRLRAEIDNVKKQCANLQNAIADAEQRGELALKDARNKLAELEEALQKAKQDMARLLREYQELMNTKLALDVEIATYRKLLEGEECRLSGEGVGPVNISVVTGSVSSGYGSGSGYGGGLGGGLGGGLGGGLAGGGSGSYYSSSSGGVGLSGGLSVGGSGFSASSGRGLGVGFGSGGGSSSSVKFVSTTSSSRKSFKS
Specifications
AAH24292 | |
keratin 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT5 | |
GST |
3852 | |
Wheat germ expression system | |
10 μg | |
CK5, DDD, EBS2, K5, KRT5A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |