missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRTHB1 (Human) Recombinant Protein
Human KRTHB1 full-length ORF ( AAH21241, 1 a.a. - 202 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003887-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKATVIRHGETLRRTKEEINELNRMIQRLTAEVENAKCQNSKLEAAVAQSEQQGEAALSDARCKLAELEGALQKAKQDMACLIREYQEVMNSKLGLDIEIATYRRLLEGEEQRLCEGIGAVNVCVSSSRGGVVCGDLCVSGSRPVTGSVCSAPCNGNVAVSTGLCAPCGQLNTTCGGGSCGVGSCGISSLGVGSCGSSCRKC
Specifications
AAH21241 | |
keratin 81 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT81 | |
GST |
3887 | |
Wheat germ expression system | |
25 μg | |
HB1, Hb-1, KRTHB1, MLN137, ghHkb1, hHAKB2-1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |