missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
STEMCELL Technologies Ku-0063794
Beskrivning
- Ku-0063794 is a cell-permeable, selective inhibitor of the serine-threonine kinase mammalian target of rapamycin (mTOR), inhibiting both the mTORC1 and mTORC2 complexes (IC50 = 10 nM)
- It shows good specificity (> 1000-fold) against 76 other protein kinases or 7 lipid kinases, including PI3 kinases (García-Martínez et al.)
- Extends the lifespan of Toll-like receptor (TLR)-activated dendritic cells by preserving mitochondrial oxidative phosphorylation (Amiel et al.)
- Inhibits cell growth by inducing G1-cell cycle arrest in mouse embryonic fibroblasts and human non-small cell lung carcinoma cell lines (García-Martínez et al.; Fei et al.)
- Inhibits tumor growth in a xenograft model of renal cell carcinoma (Zhang et al.)
- Reduces keloid (fibroproliferative dermal lesion) volume in an ex vivo keloid organ culture model, and inhibits keloid cell spreading, proliferation, migration, and invasive properties in vitro (Syed et al.)
Specifikationer
Specifikationer
| CAS | 938440-64-3 |
| Kvantitet | 5 mg |
| Molekylformel | C25H31N5O4 |
| Renhet | ≥98% |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?