Läs mer
Tocris Bioscience™ KW 3902
GREENER_CHOICE
Produktkod. 18003862
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18003862 10 mg
18004262 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18003862 Leverantör Tocris Bioscience™ Leverantörsnummer 4167/10
Selective A1 antagonist
Selective adenosine A1 receptor antagonist; displays 890-fold selectivity for rat A1 receptors over A2A receptors (Ki values are 0.19 and 170 nM respectively). Displays no effect on recombinant rat A3 receptors expressed on CHO cells at concentrations up to 10 μM. Exhibits diuretic and renal protective effects in rats.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136199-02-5
Molekylformel C20H28N4O2
Specifikationer
Kemiskt namn eller material KW 3902
Kvantitet 10 mg
Formel vikt 356.46
Renhet >99%
Molekylvikt (g/mol) 356.47