missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ KW 3902
GREENER_CHOICE

Produktkod. 18004262
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18004262 50 mg
18003862 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18004262 missing translation for 'mfr' Tocris Bioscience™ missing translation for 'supplierNo' 4167/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective A1 antagonist

Selective adenosine A1 receptor antagonist; displays 890-fold selectivity for rat A1 receptors over A2A receptors (Ki values are 0.19 and 170 nM respectively). Displays no effect on recombinant rat A3 receptors expressed on CHO cells at concentrations up to 10 μM. Exhibits diuretic and renal protective effects in rats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136199-02-5
Molekylformel C20H28N4O2

Specifications

Kemiskt namn eller material KW 3902
Kvantitet 50 mg
Formel vikt 356.46
Renhet >99%
Molekylvikt (g/mol) 356.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.