Läs mer
Tocris Bioscience™ KYT 0353
Beskrivning
KYT 0353 is a potent and selective inhibitor of L-type amino acid transporter 1 (LAT1) (IC50 = 60 nM for 14C-leucine uptake in H-29 cells). Also suppreses 14C-leucine uptake in renal proximal tubule cells expressing LAT1 (IC50 = 140 nM). Displays ∽500-fold selectivity for LAT1 over LAT2. Reduces cell viability and induces apoptosis in YD-38 oral cancer cells. Arrests growth in several tumor cell lines. Exhibits growth inhibitory effects on HT-29 cells in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | O-[(5-Amino-2-phenyl-7-benzoxazolyl)methyl]-3,5-dichloro-L-tyrosine dihydrochloride |
| CAS | 1597402-27-1 |
| Synonym | JPH 203 |
| Kvantitet | 5 mg |
| Molekylformel | C23H19Cl2N3O4.2HCl |
| Formel vikt | 545.24 |
| Produkttyp | Inhibitor |
| Renhet | 96% |