Läs mer
Tocris Bioscience™ L 006235
GREENER_CHOICE
Produktkod. 18084642
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18084642 50 mg
18055082 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18084642 Leverantör Tocris Bioscience™ Leverantörsnummer 3066/50
Potent cathepsin K inhibitor
Potent, reversible cathepsin K inhibitor (IC50 = 0.25 nM) that displays > 4000-fold selectivity over cathepsins B, L and S. Displays reduced selectivity in cell-based assays possibly due to lysosomal accumulation. Reduces collagen breakdown and promotes bone deposition in vivo. Orally active and has intrinsic fluorescence.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 294623-49-7
Molekylformel C24H30N6O2S
Specifikationer
Kemiskt namn eller material L 006235
Kvantitet 50 mg
Formel vikt 466.6
Renhet >98%
Molekylvikt (g/mol) 466.604