Läs mer
Tocris Bioscience™ L-152,804
GREENER_CHOICE
Produktkod. 18085862
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18085862 10 mg
18034922 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18085862 Leverantör Tocris Bioscience™ Leverantörsnummer 1382/10
Potent, selective non-peptide NPY Y5 receptor antagonist
Potent, selective non-peptide neuropeptide Y Y5 receptor antagonist (Ki = 26 nM for hY5). Displays > 300-fold selectivity over hY1, hY2, and hY4 receptors. Causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure. Centrally active upon oral administration in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6508-43-6
Molekylformel C23H26O4
Specifikationer
Kemiskt namn eller material L-152, 804
Kvantitet 10 mg
Formel vikt 366.46
Renhet >99%
Molekylvikt (g/mol) 366.457