Läs mer
L-2-Amino-3-[thiomethyl-1-(1-oxyl-2,2,5,5-tetramethyl-3 -pyrrolin-3-yl)]propanoic Acid Dihydrochloride, TRC
Produktkod. 30324697
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30324697 5 mg
1 options
Produktkod. 30324697 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6304005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-2-Amino-3-[thiomethyl-1-(1-oxyl-2,2,5,5-tetramethyl-3 -pyrrolin-3-yl)]propanoic Acid Dihydrochloride
CAS 1356577-86-0
Molekylformel C12H21N2O3S.. 2(HCl)
Kvantitet 5 mg
Synonym 3-[[[(2R)-2-Amino-2-carboxyethyl]thio]methyl]-2,5-dihydro-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy Dihydrochloride
InChI formel InChI=1S/C12H21N2O3S.2ClH/c1-11(2)5-8(12(3,4)14(11)17)6-18-7-9(13)10(15)16;;/h5,9H,6-7,13H2,1-4H3,(H,15,16);2*1H/t9-;;/m0../s1
LEDER N[C@H](C(O)=O)CSCC1=CC(C)(C)N([O])C1(C)C.Cl.Cl
IUPAC-namn (2R)-2-amino-3-[(1-hydroxy-2,2,5,5-tetramethylpyrrol-3-yl)methylsulfanyl]propanoic acid;dihydrochloride
Molekylvikt (g/mol) 346.29
Formel vikt 345.09