Läs mer
(L)-2-Bromo-3-phenylpropionic Acid, TRC
Produktkod. 30340884
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30340884 2.5 g
1 options
Produktkod. 30340884 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6864702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (L)-2-Bromo-3-phenylpropionic Acid
CAS 35016-63-8
Molekylformel C9 H9 Br O2
Kvantitet 2.5 g
Synonym Benzenepropanoic acid, α-bromo-, (αS)-, Benzenepropanoic acid, α-bromo-, (S)-, (αS)-α-Bromobenzenepropanoic acid, (S)-α-Bromobenzenepropanoic acid, 3-Phenyl-2(S)-bromopropionic acid, L-α-Bromo-β-phenylpropionic acid, (S)-2-Bromo-3-phenylpropionic Acid
InChI formel InChI=1S/C9H9BrO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6H2,(H,11,12)/t8-/m0/s1
LEDER OC(=O)[C@@H](Br)Cc1ccccc1
IUPAC-namn (2S)-2-bromo-3-phenylpropanoic acid
Molekylvikt (g/mol) 229.07
Formel vikt 227.9786