Läs mer
Tocris Bioscience™ L-689,560
Highly potent NMDA antagonist
Brand: Tocris Bioscience™ 0742/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
L-689,560 is a very potent antagonist at the glycine-NMDA site.Specifikationer
| trans-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline | |
| >99% | |
| Soluble to 25mM in DMSO |
| 139051-78-8 | |
| 10 mg |
Produkttitel
For Research Use Only