missing translation for 'onlineSavingsMsg'
Läs mer

L-732,138, Tocris Bioscience™

Produktkod. 18706457
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18706457 10 mg
18054282 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18706457 Leverantör Tocris Bioscience Leverantörsnummer 0868/10

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Potent and selective NK1 antagonist

Potent and highly selective competitive tachykinin NK1 receptor antagonist (IC50 = 2.3 nM).
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 148451-96-1
Molekylformel C22H18F6N2O3
Molekylvikt (g/mol) 472.387
InChI-nyckel BYYQYXVAWXAYQC-IBGZPJMESA-N
Synonym 3,5-bis tfm bz nactrp, 3,5-bis trifluoromethyl benzyl n-acetyltryptophan, n-acetyltryptophan 3,5-bis trifluoromethyl benzyl ester, chembl22870, n-acetyl-l-trp-3,5-bistrifluoromethylbenzyl ester, 3,5-bis trifluoromethyl phenyl methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, n-acetyl-l-tryptophan-3,5-bistrifluoromethylbenzyl ester, n-acetyl-l-tryptophan 3,5-bis trifluoromethyl benzyl ester, l-tryptophan, n-acetyl-, 3,5-bis trifluoromethyl phenyl methyl ester, tocris-0868
PubChem CID 132837
IUPAC-namn [3,5-bis(trifluoromethyl)phenyl]methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
LEDER CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)OCC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F

Specifikationer

Kemiskt namn eller material L-732, 138
Kvantitet 10 mg
Formel vikt 472.39
Procent renhet >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.