Läs mer
L-a-Acetoxypropionyl Chloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-alpha-Acetoxypropionyl Chloride |
| CAS | 36394-75-9 |
| Molekylformel | C5H7ClO3 |
| Kvantitet | 5 g |
| Synonym | (2S)-2-(Acetyloxy)propanoyl Chloride, (1S)-2-Chloro-1-methyl-2-oxoethyl Acetate, (2S)-1-Chloro-1-oxopropan-2-yl Acetate, (2S)-2-Acetoxypropionyl Chloride, (S)-(-)-2-Acetoxypropionyl Chloride, (S)-1-Chloro-1-oxopropan-2-yl Acetate, (S)-2-(Acetyloxy)propanoyl Chloride, (S)-2-Acetoxypropanoyl Chloride, (S)-2-Acetoxypropionoyl Chloride, (S)-2-Acetoxypropionyl Chloride, (S)-2-Acetyloxypropionyl Chloride, 2-(S)-Acetoxypropanoyl Chloride, Acetic Acid (1S)-1-(Chlorocarbonyl)ethyl Ester, Acetic Acid (1S)-2-Chloro-1-methyl-2-oxoethyl Ester, Acetic Acid (2S)-1-Chloro-1-oxopropan-2-yl Ester |
| InChI formel | InChI=1S/C5H7ClO3/c1-3(5(6)8)9-4(2)7/h3H,1-2H3/t3-/m0/s1 |
| LEDER | C[C@H](OC(=O)C)C(=O)Cl |
| IUPAC-namn | [(2S)-1-chloro-1-oxopropan-2-yl] acetate |
| Molekylvikt (g/mol) | 150.56 |
| Formel vikt | 150.0084 |